missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dibromomaleimide 98.0+%, TCI America™

Catalog No. D39101G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39101G 1 g
D39105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39101G Supplier TCI America Supplier No. D39101G

Chemical Identifiers

CAS 1122-10-7
Molecular Formula C4HBr2NO2
Molecular Weight (g/mol) 254.865
MDL Number MFCD00185696
InChI Key BIKSKRPHKQWJCW-UHFFFAOYSA-N
PubChem CID 14279
IUPAC Name 3,4-dibromopyrrole-2,5-dione
SMILES C1(=C(C(=O)NC1=O)Br)Br

Specifications

Melting Point 228°C
Quantity 1 g
UN Number 1759
Formula Weight 254.87
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dibromomaleimide
RTECSNumber ON5100000
TSCA No