missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dibenzyloxybenzaldehyde 98.0+%, TCI America™
Supplier: TCI America D407925G
Specifications
| 3,4-Dibenzyloxybenzaldehyde | |
| 92°C | |
| MFCD00004776 | |
| 3,4-Bis(benzyloxy)benzaldehyde | |
| C1=CC=C(C=C1)COC2=C(C=C(C=C2)C=O)OCC3=CC=CC=C3 | |
| 318.37 | |
| 318.37 | |
| Crystalline Powder |
| 5447-02-9 | |
| C21H18O3 | |
| 25 g | |
| XDDLXZHBWVFPRG-UHFFFAOYSA-N | |
| 3,4-bis(phenylmethoxy)benzaldehyde | |
| 79526 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 5447-02-9 | |
| 318.37 | |
| XDDLXZHBWVFPRG-UHFFFAOYSA-N | |
| 79526 | |
| C1=CC=C(C=C1)COC2=C(C=C(C=C2)C=O)OCC3=CC=CC=C3 |
| C21H18O3 | |
| MFCD00004776 | |
| 3,4-Bis(benzyloxy)benzaldehyde | |
| 3,4-bis(phenylmethoxy)benzaldehyde |
Safety and Handling
TSCA : No