missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dibenzyloxybenzaldehyde 98.0+%, TCI America™

Catalog No. p-7128087
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40795G 5 g
D407925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40795G Supplier TCI America Supplier No. D40795G

Chemical Identifiers

CAS 5447-02-9
Molecular Formula C21H18O3
Molecular Weight (g/mol) 318.37
MDL Number MFCD00004776
InChI Key XDDLXZHBWVFPRG-UHFFFAOYSA-N
Synonym 3,4-Bis(benzyloxy)benzaldehyde
PubChem CID 79526
IUPAC Name 3,4-bis(phenylmethoxy)benzaldehyde
SMILES C1=CC=C(C=C1)COC2=C(C=C(C=C2)C=O)OCC3=CC=CC=C3

Specifications

Melting Point 92°C
Quantity 5 g
Formula Weight 318.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dibenzyloxybenzaldehyde