missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Diaminotoluene 98.0+%, TCI America™
$164.03 - $164.03
Chemical Identifiers
| CAS | 496-72-0 |
|---|---|
| Molecular Formula | C7H10N2 |
| Molecular Weight (g/mol) | 122.171 |
| MDL Number | MFCD00007728 |
| InChI Key | DGRGLKZMKWPMOH-UHFFFAOYSA-N |
| Synonym | 3,4-diaminotoluene, 3,4-toluenediamine, 1,2-benzenediamine, 4-methyl, 4-methyl-o-phenylenediamine, toluene-3,4-diamine, 4-methyl-1,2-phenylenediamine, 4-methyl-1,2-benzenediamine, 3,4-tolylenediamine, 3,4-toluylenediamine, 1,2-diamino-4-methylbenzene |
| PubChem CID | 10332 |
| IUPAC Name | 4-methylbenzene-1,2-diamine |
| SMILES | CC1=CC(=C(C=C1)N)N |
Chemical Identifiers
| 496-72-0 | |
| 122.171 | |
| DGRGLKZMKWPMOH-UHFFFAOYSA-N | |
| 10332 | |
| CC1=CC(=C(C=C1)N)N |
| C7H10N2 | |
| MFCD00007728 | |
| 3,4-diaminotoluene, 3,4-toluenediamine, 1,2-benzenediamine, 4-methyl, 4-methyl-o-phenylenediamine, toluene-3,4-diamine, 4-methyl-1,2-phenylenediamine, 4-methyl-1,2-benzenediamine, 3,4-tolylenediamine, 3,4-toluylenediamine, 1,2-diamino-4-methylbenzene | |
| 4-methylbenzene-1,2-diamine |
Specifications
| 496-72-0 | |
| 265°C | |
| MFCD00007728 | |
| 3,4-diaminotoluene, 3,4-toluenediamine, 1,2-benzenediamine, 4-methyl, 4-methyl-o-phenylenediamine, toluene-3,4-diamine, 4-methyl-1,2-phenylenediamine, 4-methyl-1,2-benzenediamine, 3,4-tolylenediamine, 3,4-toluylenediamine, 1,2-diamino-4-methylbenzene | |
| CC1=CC(=C(C=C1)N)N | |
| 122.171 | |
| 122.17 | |
| Crystalline Lumps |
| 90°C | |
| C7H10N2 | |
| 25 g | |
| DGRGLKZMKWPMOH-UHFFFAOYSA-N | |
| 4-methylbenzene-1,2-diamine | |
| 10332 | |
| ≥98.0% (T) | |
| 3,4-Diaminotoluene |
Safety and Handling
EINECSNumber : (3)-0126
RTECSNumber : XS9820000
TSCA : Yes