missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Diaminotoluene 98.0+%, TCI America™
Supplier: TCI America D012525G
Specifications
| 3,4-Diaminotoluene | |
| 90°C | |
| C7H10N2 | |
| 25 g | |
| DGRGLKZMKWPMOH-UHFFFAOYSA-N | |
| 4-methylbenzene-1,2-diamine | |
| 10332 | |
| ≥98.0% (T) |
| 496-72-0 | |
| 265°C | |
| MFCD00007728 | |
| 3,4-diaminotoluene, 3,4-toluenediamine, 1,2-benzenediamine, 4-methyl, 4-methyl-o-phenylenediamine, toluene-3,4-diamine, 4-methyl-1,2-phenylenediamine, 4-methyl-1,2-benzenediamine, 3,4-tolylenediamine, 3,4-toluylenediamine, 1,2-diamino-4-methylbenzene | |
| CC1=CC(=C(C=C1)N)N | |
| 122.171 | |
| 122.17 | |
| Crystalline Lumps |
Chemical Identifiers
| 496-72-0 | |
| 122.171 | |
| DGRGLKZMKWPMOH-UHFFFAOYSA-N | |
| 10332 | |
| CC1=CC(=C(C=C1)N)N |
| C7H10N2 | |
| MFCD00007728 | |
| 3,4-diaminotoluene, 3,4-toluenediamine, 1,2-benzenediamine, 4-methyl, 4-methyl-o-phenylenediamine, toluene-3,4-diamine, 4-methyl-1,2-phenylenediamine, 4-methyl-1,2-benzenediamine, 3,4-tolylenediamine, 3,4-toluylenediamine, 1,2-diamino-4-methylbenzene | |
| 4-methylbenzene-1,2-diamine |
Safety and Handling
EINECSNumber : (3)-0126
RTECSNumber : XS9820000
TSCA : Yes