missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminothiophene Dihydrochloride 98.0+%, TCI America™

Catalog No. D23781G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23781G 1 g
D2378100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23781G Supplier TCI America Supplier No. D23781G

Chemical Identifiers

CAS 90069-81-1
Molecular Formula C4H8Cl2N2S
Molecular Weight (g/mol) 187.082
MDL Number MFCD00671511
InChI Key RAMOMCXNLLLICQ-UHFFFAOYSA-N
Synonym 3,4-diaminothiophene dihydrochloride, thiophene-3,4-diamine dihydrochloride, 3,4-diaminothiophenedihydrochloride, pubchem21822, acmc-209r4q, 3,4-diaminothiophene 2hcl, 3?4-diaminothiophene dihydrochloride, 3,4-thiophenediamine, dihydrochloride, thiophene-3,4-diamine-hydrogen chloride 1/2
PubChem CID 12707342
IUPAC Name thiophene-3,4-diamine;dihydrochloride
SMILES C1=C(C(=CS1)N)N.Cl.Cl

Specifications

Color Yellow
Quantity 1 g
Formula Weight 187.08
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Diaminothiophene Dihydrochloride
TSCA No
Recommended Storage Refrigerator