missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminopyridine, 97%

CAS: 54-96-6 | C5H7N3 | 109.132 g/mol

$84.38 - $1006.22

Chemical Identifiers

CAS 54-96-6
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.132
MDL Number MFCD00006401
InChI Key OYTKINVCDFNREN-UHFFFAOYSA-N
Synonym 3,4-diaminopyridine, amifampridine, 3,4-pyridinediamine, firdapse, diamino-3,4 pyridine, 4,5-diaminopyridine, 3,4-dap, amifampridin, zenas, pyridine, 3,4-diamino
PubChem CID 5918
IUPAC Name pyridine-3,4-diamine
SMILES C1=CN=CC(=C1N)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1246503
View Documents
Thermo Scientific Chemicals
A1246503
1 g
Each for $84.38
Only null left
 
AAA1246506
View Documents
Thermo Scientific Chemicals
A1246506
5 g
Each for $270.10
Only null left
 
AAA1246514
View Documents
Thermo Scientific Chemicals
A1246514
25 g
Each for $1,006.22
Only null left
 
Description

Description

It is a potassium channel blocker in nerve terminals. It inhibits potassium channel efflux, increasing the duration of the action potential, which results in an increase in the duration of calcium channel opening and enhanced acetylcholine (ACh) release. Increased ACh availability at the motor end plate allows muscles to contract.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a potassium channel blocker in nerve terminals. It inhibits potassium channel efflux, increasing the duration of the action potential, which results in an increase in the duration of calcium channel opening and enhanced acetylcholine (ACh) release. Increased ACh availability at the motor end plate allows muscles to contract.

Solubility
Soluble in water (24 mg/ml at 20°C), alcohol. Slightly soluble in ether.

Notes
Store under cool dry place. Ensure good ventilation. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

54-96-6
109.132
OYTKINVCDFNREN-UHFFFAOYSA-N
5918
C1=CN=CC(=C1N)N
C5H7N3
MFCD00006401
3,4-diaminopyridine, amifampridine, 3,4-pyridinediamine, firdapse, diamino-3,4 pyridine, 4,5-diaminopyridine, 3,4-dap, amifampridin, zenas, pyridine, 3,4-diamino
pyridine-3,4-diamine

Specifications

54-96-6
240°C (464°F)
C5H7N3
1 g
110232
3,4-diaminopyridine, amifampridine, 3,4-pyridinediamine, firdapse, diamino-3,4 pyridine, 4,5-diaminopyridine, 3,4-dap, amifampridin, zenas, pyridine, 3,4-diamino
OYTKINVCDFNREN-UHFFFAOYSA-N
pyridine-3,4-diamine
5918
≥98%
217°C to 220°C
Unpleasant
MFCD00006401
UN2811
14,2986
Soluble in water (24mg/ml at 20°C),alcohol. Slightly soluble in ether.
C1=CN=CC(=C1N)N
109.132
109.13
3,4-Diaminopyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H312-H319
Fatal if swallowed.
Fatal if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.

P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c

H300+H330-H311-H315-H318-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 200-220-9

RTECSNumber : US7600000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only