missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminopyridine, 97%

CAS: 54-96-6 | C5H7N3 | 109.132 g/mol

Supplier:  Thermo Scientific Chemicals A1246506

 View more versions of this product

Catalog No. AAA1246506


Only null left
Explore available promotions

Description

Description

It is a potassium channel blocker in nerve terminals. It inhibits potassium channel efflux, increasing the duration of the action potential, which results in an increase in the duration of calcium channel opening and enhanced acetylcholine (ACh) release. Increased ACh availability at the motor end plate allows muscles to contract.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a potassium channel blocker in nerve terminals. It inhibits potassium channel efflux, increasing the duration of the action potential, which results in an increase in the duration of calcium channel opening and enhanced acetylcholine (ACh) release. Increased ACh availability at the motor end plate allows muscles to contract.

Solubility
Soluble in water (24 mg/ml at 20°C), alcohol. Slightly soluble in ether.

Notes
Store under cool dry place. Ensure good ventilation. Incompatible with oxidizing agents.
Specifications

Specifications

3,4-Diaminopyridine
217°C to 220°C
Unpleasant
MFCD00006401
UN2811
14,2986
Soluble in water (24mg/ml at 20°C),alcohol. Slightly soluble in ether.
C1=CN=CC(=C1N)N
109.132
109.13
54-96-6
240°C (464°F)
C5H7N3
5 g
110232
3,4-diaminopyridine, amifampridine, 3,4-pyridinediamine, firdapse, diamino-3,4 pyridine, 4,5-diaminopyridine, 3,4-dap, amifampridin, zenas, pyridine, 3,4-diamino
OYTKINVCDFNREN-UHFFFAOYSA-N
pyridine-3,4-diamine
5918
≥98%

Chemical Identifiers

54-96-6
109.132
OYTKINVCDFNREN-UHFFFAOYSA-N
5918
C1=CN=CC(=C1N)N
C5H7N3
MFCD00006401
3,4-diaminopyridine, amifampridine, 3,4-pyridinediamine, firdapse, diamino-3,4 pyridine, 4,5-diaminopyridine, 3,4-dap, amifampridin, zenas, pyridine, 3,4-diamino
pyridine-3,4-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H312-H319
Fatal if swallowed.
Fatal if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.

P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c

H300+H330-H311-H315-H318-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 200-220-9

RTECSNumber : US7600000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only