missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4'-Diaminodiphenyl Ether 97.0+%, TCI America™
$123.03 - $480.92
Chemical Identifiers
| CAS | 2657-87-6 |
|---|---|
| Molecular Formula | C12H12N2O |
| Molecular Weight (g/mol) | 200.241 |
| MDL Number | MFCD00036097 |
| InChI Key | ZBMISJGHVWNWTE-UHFFFAOYSA-N |
| PubChem CID | 75871 |
| IUPAC Name | 3-(4-aminophenoxy)aniline |
| SMILES | C1=CC(=CC(=C1)OC2=CC=C(C=C2)N)N |
Chemical Identifiers
| 2657-87-6 | |
| 200.241 | |
| ZBMISJGHVWNWTE-UHFFFAOYSA-N | |
| 3-(4-aminophenoxy)aniline |
| C12H12N2O | |
| MFCD00036097 | |
| 75871 | |
| C1=CC(=CC(=C1)OC2=CC=C(C=C2)N)N |
Specifications
| 2657-87-6 | |
| C12H12N2O | |
| 5 g | |
| ZBMISJGHVWNWTE-UHFFFAOYSA-N | |
| 3-(4-aminophenoxy)aniline | |
| 75871 | |
| ≥97.0% (GC,T) | |
| 3,4′-Diaminodiphenyl Ether |
| 207°C | |
| MFCD00036097 | |
| 3,4′C-Oxydianiline | |
| C1=CC(=CC(=C1)OC2=CC=C(C=C2)N)N | |
| 200.241 | |
| 200.24 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-3633
TSCA : No