missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4'-Diaminodiphenyl Ether 97.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 207°C |
| Quantity | 5 g |
| Synonym | 3,4′C-Oxydianiline |
| Formula Weight | 200.24 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,4′-Diaminodiphenyl Ether |
Safety and Handling
| EINECSNumber | (3)-3633 |
| TSCA | No |