missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminobenzonitrile 98.0+%, TCI America™

Catalog No. D51941G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51941G 1 g
D51945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51941G Supplier TCI America Supplier No. D51941G

Chemical Identifiers

CAS 17626-40-3
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00723901
InChI Key VWLLPPSBBHDXHK-UHFFFAOYSA-N
Synonym 3,4-diamino-benzonitrile, benzonitrile, 3,4-diamino, 4-cyanobenzene-1,2-diamine, 3,4-diaminobenzenecarbonitrile, zlchem 619, pubchem2547, 3, 4-diaminobenzonitrile, 3,4 diamino benzonitrile, 3,4-diamino benzonitrile, acmc-209eb5
PubChem CID 737285
IUPAC Name 3,4-diaminobenzonitrile
SMILES NC1=C(N)C=C(C=C1)C#N

Specifications

Melting Point 149°C
Quantity 1 g
UN Number 3439
Formula Weight 133.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Diaminobenzonitrile