accessibility menu, dialog, popup

UserName

NH2NNH2

CAS RN 17626-40-3

IUPAC Name: 3,4-diaminobenzonitrile
Synonyms: 3,4-diaminobenzonitrile
Molecular Weight (g/mol): 133.15
Molecular Formula: C7H7N3
InChi Key: VWLLPPSBBHDXHK-UHFFFAOYSA-N
SMILES: NC1=C(N)C=C(C=C1)C#N
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)