missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminoanisole dihydrochloride, 98%

Catalog No. p-4812878
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC396990010 1 g
AC396990050 5 g
AC396990250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC396990010 Supplier Thermo Scientific Chemicals Supplier No. 396990010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59548-39-9 | C7H10N2O·2ClH | 211.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59548-39-9
Molecular Formula C7H10N2O·2ClH
Molecular Weight (g/mol) 211.09
InChI Key SXCHMHOBHJOXGC-UHFFFAOYSA-N
Synonym 4-methoxybenzene-1,2-diamine dihydrochloride, 4-methoxy-o-phenylenediamine dihydrochloride, 3,4-diaminoanisole dihydrochloride, 1,2-benzenediamine, 4-methoxy-, dihydrochloride, 3,4-diaminoanisole 2hcl, 4-methoxy-phenylenediamine dihydrochloride, 4-methoxy-1,2-benzenediamine dihydrochloride, 4-methoxy-benzene-1,2-diamine dihydrochloride, 4-methoxy-1,2-phenylenediamine dihydrochloride, 4-methoxy-l,2-phenylene diamine dihydrochloride
PubChem CID 3085161
IUPAC Name 4-methoxybenzene-1,2-diamine;dihydrochloride
SMILES COC1=CC(=C(C=C1)N)N.Cl.Cl

Specifications

Melting Point 206.0°C to 209.0°C
Assay Percent Range 97.5 to max. 102.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3OC6H3(NH2)2·2HCl
Quantity 1 g
Solubility Information Solubility in water: soluble.
Formula Weight 211.09
Percent Purity 98%
Chemical Name or Material 3, 4-Diaminoanisole dihydrochloride
Hazard Category GHS Signal Word: Warning
EINECSNumber 261-801-

RUO – Research Use Only