missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminoanisole dihydrochloride, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 396990050

Quantity 5g

 View more versions of this product

Catalog No. AC396990050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

59548-39-9
211.09
4-methoxybenzene-1,2-diamine dihydrochloride, 4-methoxy-o-phenylenediamine dihydrochloride, 3,4-diaminoanisole dihydrochloride, 1,2-benzenediamine, 4-methoxy-, dihydrochloride, 3,4-diaminoanisole 2hcl, 4-methoxy-phenylenediamine dihydrochloride, 4-methoxy-1,2-benzenediamine dihydrochloride, 4-methoxy-benzene-1,2-diamine dihydrochloride, 4-methoxy-1,2-phenylenediamine dihydrochloride, 4-methoxy-l,2-phenylene diamine dihydrochloride
4-methoxybenzene-1,2-diamine;dihydrochloride
C7H10N22ClH
SXCHMHOBHJOXGC-UHFFFAOYSA-N
3085161
COC1=CC(=C(C=C1)N)N.Cl.Cl

Specifications

3, 4-Diaminoanisole dihydrochloride
97.5 to max. 102.5% min. (Argentometry)
C7H10N22ClH
5g
Solubility in water: soluble.
COC1=CC(=C(C=C1)N)N.Cl.Cl
211.09
211.09
59548-39-9
Glass bottle
CH3OC6H3(NH2)2·2HCl
4-methoxybenzene-1,2-diamine dihydrochloride, 4-methoxy-o-phenylenediamine dihydrochloride, 3,4-diaminoanisole dihydrochloride, 1,2-benzenediamine, 4-methoxy-, dihydrochloride, 3,4-diaminoanisole 2hcl, 4-methoxy-phenylenediamine dihydrochloride, 4-methoxy-1,2-benzenediamine dihydrochloride, 4-methoxy-benzene-1,2-diamine dihydrochloride, 4-methoxy-1,2-phenylenediamine dihydrochloride, 4-methoxy-l,2-phenylene diamine dihydrochloride
SXCHMHOBHJOXGC-UHFFFAOYSA-N
4-methoxybenzene-1,2-diamine;dihydrochloride
3085161
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 261-801-

SDS
Documents

Documents

Promotions

Promotions