missing translation for 'onlineSavingsMsg'
Learn More

3,4'-Diamino-4-nitrodiphenyl Ether 98.0+%, TCI America™

Catalog No. D36755G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D36755G Supplier TCI America Supplier No. D36755G
Only null left

Chemical Identifiers

CAS 30491-74-8
Molecular Formula C12H11N3O3
Molecular Weight (g/mol) 245.24
MDL Number MFCD15072156
InChI Key ZHELWQKSRPWTPN-UHFFFAOYSA-N
PubChem CID 13705242
IUPAC Name 5-(4-aminophenoxy)-2-nitroaniline
SMILES NC1=CC=C(OC2=CC(N)=C(C=C2)[N+]([O-])=O)C=C1

Specifications

Melting Point 170°C
Quantity 5 g
Formula Weight 245.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4′-Diamino-4-nitrodiphenyl Ether