missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diacetyl-2,5-hexanedione 98.0+%, TCI America™

Supplier:  TCI America D4826200MG

 View more versions of this product

Catalog No. D4826200MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3,4-Diacetyl-2,5-hexanedione
193°C
122°C
MFCD00031531
1,1,2,2-Tetraacetylethane, Bis(acetylacetone)
CC(=O)C(C(C(C)=O)C(C)=O)C(C)=O
198.22
198.22
Crystalline Powder
5027-32-7
White-Yellow
C10H14O4
200 mg
CSKRBHOAJUMOKJ-UHFFFAOYSA-N
3,4-diacetylhexane-2,5-dione
78730
≥98.0% (GC)

Chemical Identifiers

5027-32-7
198.22
CSKRBHOAJUMOKJ-UHFFFAOYSA-N
78730
CC(=O)C(C(C(C)=O)C(C)=O)C(C)=O
C10H14O4
MFCD00031531
1,1,2,2-Tetraacetylethane, Bis(acetylacetone)
3,4-diacetylhexane-2,5-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions