missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Diacetyl-2,5-hexanedione 98.0+%, TCI America™
Supplier: TCI America D48261G
Specifications
| 3,4-Diacetyl-2,5-hexanedione | |
| 193°C | |
| 122°C | |
| MFCD00031531 | |
| 1,1,2,2-Tetraacetylethane, Bis(acetylacetone) | |
| CC(=O)C(C(C(C)=O)C(C)=O)C(C)=O | |
| 198.22 | |
| 198.22 | |
| Crystalline Powder |
| 5027-32-7 | |
| White-Yellow | |
| C10H14O4 | |
| 1 g | |
| CSKRBHOAJUMOKJ-UHFFFAOYSA-N | |
| 3,4-diacetylhexane-2,5-dione | |
| 78730 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 5027-32-7 | |
| 198.22 | |
| CSKRBHOAJUMOKJ-UHFFFAOYSA-N | |
| 78730 | |
| CC(=O)C(C(C(C)=O)C(C)=O)C(C)=O |
| C10H14O4 | |
| MFCD00031531 | |
| 1,1,2,2-Tetraacetylethane, Bis(acetylacetone) | |
| 3,4-diacetylhexane-2,5-dione |
Safety and Handling
TSCA : No