missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Diacetoxybenzaldehyde 98.0+%, TCI America™
Supplier: TCI America D20425G
Specifications
| 3,4-Diacetoxybenzaldehyde | |
| 53°C | |
| C11H10O5 | |
| 5 g | |
| CC(=O)OC1=C(OC(C)=O)C=C(C=O)C=C1 | |
| 222.20 | |
| 222.20 | |
| Crystalline Powder |
| 67727-64-4 | |
| White-Yellow | |
| MFCD00143057 | |
| WYHMNJKAVNPOOR-UHFFFAOYSA-N | |
| 2-(acetyloxy)-5-formylphenyl acetate | |
| 1517488 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 67727-64-4 | |
| 222.20 | |
| WYHMNJKAVNPOOR-UHFFFAOYSA-N | |
| 2-(acetyloxy)-5-formylphenyl acetate |
| C11H10O5 | |
| MFCD00143057 | |
| 1517488 | |
| CC(=O)OC1=C(OC(C)=O)C=C(C=O)C=C1 |
Safety and Handling
TSCA : No