missing translation for 'onlineSavingsMsg'
Learn More

3,4-Di-n-butoxy-3-cyclobutene-1,2-dione, 97%

Catalog No. aaa1896014
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1896014 25 g
AAA1896003 1 g
AAA1896006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1896014 Supplier Thermo Scientific Chemicals Supplier No. A1896014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2892-62-8 | C12H18O4 | 226.27 g/mol

3,4-Dibutoxy-3-cyclobutene-1,2-dione was used in the synthesis of novel semisquarylium dye, useful for highly selective Hg2+ sensing in aqueous media. It was also employed in the synthesis of 3-butoxy-4-(1,3,3-trimethyl-2,3-dihydro-1H-2-indolylidenemethyl)-3-cyclobutene-1,2-dione, intermediate for the synthesis of squaryl dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dibutoxy-3-cyclobutene-1,2-dione was used in the synthesis of novel semisquarylium dye, useful for highly selective Hg2+ sensing in aqueous media. It was also employed in the synthesis of 3-butoxy-4-(1,3,3-trimethyl-2,3-dihydro-1H-2-indolylidenemethyl)-3-cyclobutene-1,2-dione, intermediate for the synthesis of squaryl dyes.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 2892-62-8
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.27
MDL Number MFCD00037150
InChI Key XBRWELTXMQSEIN-UHFFFAOYSA-N
Synonym dibutyl squarate, 3,4-dibutoxy-3-cyclobutene-1,2-dione, squaric acid dibutyl ester, sadbe, squaric acid dibutylester, 3,4-di-n-butoxy-3-cyclobutene-1,2-dione, unii-4rto57vg65, 1,2-dibutyl squarate, 3-cyclobutene-1,2-dione, 3,4-dibutoxy, acmc-209h5p
PubChem CID 65108
ChEBI CHEBI:53612
IUPAC Name 3,4-dibutoxycyclobut-3-ene-1,2-dione
SMILES CCCCOC1=C(OCCCC)C(=O)C1=O

Specifications

Density 1.047
Boiling Point 155°C to 158°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.495
Quantity 25 g
Beilstein 1968189
Solubility Information Insoluble in water.
Formula Weight 226.27
Percent Purity 98%
Chemical Name or Material 3,4-Di-n-butoxy-3-cyclobutene-1,2-dione
Hazard Category H315-H317-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c
RTECSNumber GU1772066
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only