missing translation for 'onlineSavingsMsg'
Learn More

3,4-Di-n-butoxy-3-cyclobutene-1,2-dione, 97%

CAS: 2892-62-8 | C12H18O4 | 226.27 g/mol

Supplier:  Thermo Scientific Chemicals A1896003

 View more versions of this product

Catalog No. AAA1896003


Only null left
Explore available promotions

Description

Description

3,4-Dibutoxy-3-cyclobutene-1,2-dione was used in the synthesis of novel semisquarylium dye, useful for highly selective Hg2+ sensing in aqueous media. It was also employed in the synthesis of 3-butoxy-4-(1,3,3-trimethyl-2,3-dihydro-1H-2-indolylidenemethyl)-3-cyclobutene-1,2-dione, intermediate for the synthesis of squaryl dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dibutoxy-3-cyclobutene-1,2-dione was used in the synthesis of novel semisquarylium dye, useful for highly selective Hg2+ sensing in aqueous media. It was also employed in the synthesis of 3-butoxy-4-(1,3,3-trimethyl-2,3-dihydro-1H-2-indolylidenemethyl)-3-cyclobutene-1,2-dione, intermediate for the synthesis of squaryl dyes.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

3,4-Di-n-butoxy-3-cyclobutene-1,2-dione
1.047
>110°C (230°F)
1.495
1 g
dibutyl squarate, 3,4-dibutoxy-3-cyclobutene-1,2-dione, squaric acid dibutyl ester, sadbe, squaric acid dibutylester, 3,4-di-n-butoxy-3-cyclobutene-1,2-dione, unii-4rto57vg65, 1,2-dibutyl squarate, 3-cyclobutene-1,2-dione, 3,4-dibutoxy, acmc-209h5p
XBRWELTXMQSEIN-UHFFFAOYSA-N
3,4-dibutoxycyclobut-3-ene-1,2-dione
65108
226.27
2892-62-8
155°C to 158°C (3 mmHg)
C12H18O4
MFCD00037150
1968189
Insoluble in water.
CCCCOC1=C(OCCCC)C(=O)C1=O
226.27
CHEBI:53612
98%

Chemical Identifiers

2892-62-8
226.27
XBRWELTXMQSEIN-UHFFFAOYSA-N
65108
3,4-dibutoxycyclobut-3-ene-1,2-dione
C12H18O4
MFCD00037150
dibutyl squarate, 3,4-dibutoxy-3-cyclobutene-1,2-dione, squaric acid dibutyl ester, sadbe, squaric acid dibutylester, 3,4-di-n-butoxy-3-cyclobutene-1,2-dione, unii-4rto57vg65, 1,2-dibutyl squarate, 3-cyclobutene-1,2-dione, 3,4-dibutoxy, acmc-209h5p
CHEBI:53612
CCCCOC1=C(OCCCC)C(=O)C1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c

H315-H317-H319

RTECSNumber : GU1772066

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only