missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorophenyl)-1,1-dimethylurea 95.0+%, TCI America™

Catalog No. p-7124313
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C025425G 25 g
C0254500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C025425G Supplier TCI America Supplier No. C025425G
Only null left

Chemical Identifiers

CAS 150-68-5
Molecular Formula C9H11ClN2O
Molecular Weight (g/mol) 198.65
MDL Number MFCD00018556
InChI Key BMLIZLVNXIYGCK-UHFFFAOYSA-N
Synonym monuron, 3-4-chlorophenyl-1,1-dimethylurea, chlorfenidim, lirobetarex, monurex, monurox, monuruon, monuuron, telvar, herbicides, monuron
PubChem CID 8800
ChEBI CHEBI:38214
IUPAC Name 1-(4-chlorophenyl)-3,3-dimethylurea
SMILES CN(C)C(=O)NC1=CC=C(Cl)C=C1

Specifications

Melting Point 173°C
Color White
Quantity 25 g
UN Number 3077
Formula Weight 198.65
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Chlorophenyl)-1,1-dimethylurea
EINECSNumber (3)-2194
RTECSNumber YS6300000
TSCA Yes