missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorobutyl)indole-5-carbonitrile 97.0+%, TCI America™

Catalog No. p-7125819
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30861G 1 g
C30865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30861G Supplier TCI America Supplier No. C30861G

Chemical Identifiers

CAS 143612-79-7
Molecular Formula C13H13ClN2
Molecular Weight (g/mol) 232.711
MDL Number MFCD09833261
InChI Key NJJWMEJWFYRORL-UHFFFAOYSA-N
Synonym 3-(4-Chlorobutyl)-5-cyanoindole
PubChem CID 9881123
IUPAC Name 3-(4-chlorobutyl)-1H-indole-5-carbonitrile
SMILES C1=CC2=C(C=C1C#N)C(=CN2)CCCCCl

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 1 g
Formula Weight 232.71
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Chlorobutyl)indole-5-carbonitrile