accessibility menu, dialog, popup

UserName

CAS RN 143612-79-7

IUPAC Name: 3-(4-chlorobutyl)-1H-indole-5-carbonitrile
Synonyms: 3-(4-chlorobutyl)-1H-indole-5-carbonitrile
Molecular Weight (g/mol): 232.71
Molecular Formula: C13H13ClN2
InChi Key: NJJWMEJWFYRORL-UHFFFAOYSA-N
SMILES: ClCCCCC1=CNC2=CC=C(C=C12)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results