missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-S-(1-oxido-2-pyridyl)thiuronium hexafluorophosphate, 97%, Thermo Scientific™

Catalog No. AC440260050
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC440260050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC440260050 Supplier Thermo Scientific Chemicals Supplier No. 440260050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Coupling reagent

Chemical Identifiers

CAS 212333-72-7
Molecular Formula C10H16N3OS·F6P
Molecular Weight (g/mol) 371.29
InChI Key UCOGEMMJHLHOAW-UHFFFAOYSA-N
Synonym hott, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate, hott n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronium hexafluorophosphate, ambotzrl-1156, acmc-209fij, ksc911c8f, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuron, n,n,n',n'-tetramethyl-s-1-oxido-2-pyridyl thiuronhexafluorophosphate, n,n,n,n-tetramethyl-s-1-oxido-2-pyridyl thiouronium hexafluorophosphate
PubChem CID 10571248
IUPAC Name [dimethylamino-(1-oxidopyridin-1-ium-2-yl)sulfanylmethylidene]-dimethylazanium;hexafluorophosphate
SMILES CN(C)C(=[N+](C)C)SC1=CC=CC=[N+]1[O-].F[P-](F)(F)(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Elemental Analysis 30.41 to 34.29% C, 10.64 to 12.00% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 5g
Formula Weight 371.29
Percent Purity 97%
Chemical Name or Material N,N,N',N'-Tetramethyl-S-(1-oxido-2-pyridyl)thiuronium hexafluorophosphate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only