missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromophenyl)-1-propanol 98.0+%, TCI America™

Catalog No. B46571G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46571G 1 g
B46575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46571G Supplier TCI America Supplier No. B46571G

Chemical Identifiers

CAS 25574-11-2
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD09028724
InChI Key WODKXGCVVOOEIJ-UHFFFAOYSA-N
Synonym 3-4-bromophenyl propan-1-ol, 4-bromobenzenepropanol, 3-4-bromo-phenyl-propan-1-ol, benzenepropanol, 4-bromo, 3-4-bromophenyl-1-propanol, pubchem3984, 3-4-bromophenyl propanol, 3-4-bromophenyl-propanol, amth183, 3-4-bromophenyl propyl alcohol
PubChem CID 10560614
IUPAC Name 3-(4-bromophenyl)propan-1-ol
SMILES C1=CC(=CC=C1CCCO)Br

Specifications

Color Yellow
Boiling Point 162°C
Quantity 1 g
Formula Weight 215.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(4-Bromophenyl)-1-propanol