missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromophenoxy)-6-methylpyridazine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 368869-96-9
Molecular Formula C11H9BrN2O
Molecular Weight (g/mol) 265.11
MDL Number MFCD03086135
InChI Key LZEJOINDIKGWQJ-UHFFFAOYSA-N
Synonym 3-4-bromophenoxy-6-methylpyridazine, pyridazine,3-4-bromophenoxy-6-methyl, 3-4-bromanylphenoxy-6-methyl-pyridazine, 4-bromo-1-6-methylpyridazin-3-yloxy benzene
PubChem CID 2776522
IUPAC Name 3-(4-bromophenoxy)-6-methylpyridazine
SMILES CC1=NN=C(OC2=CC=C(Br)C=C2)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC21310CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC21310DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

368869-96-9
265.11
LZEJOINDIKGWQJ-UHFFFAOYSA-N
2776522
CC1=NN=C(OC2=CC=C(Br)C=C2)C=C1
C11H9BrN2O
MFCD03086135
3-4-bromophenoxy-6-methylpyridazine, pyridazine,3-4-bromophenoxy-6-methyl, 3-4-bromanylphenoxy-6-methyl-pyridazine, 4-bromo-1-6-methylpyridazin-3-yloxy benzene
3-(4-bromophenoxy)-6-methylpyridazine

Specifications

368869-96-9
100.0
C11H9BrN2O
3-4-bromophenoxy-6-methylpyridazine, pyridazine,3-4-bromophenoxy-6-methyl, 3-4-bromanylphenoxy-6-methyl-pyridazine, 4-bromo-1-6-methylpyridazin-3-yloxy benzene
CC1=NN=C(OC2=CC=C(Br)C=C2)C=C1
265.11
265.11
3-(4-Bromophenoxy)-6-methylpyridazine
94.5
95%
MFCD03086135
LZEJOINDIKGWQJ-UHFFFAOYSA-N
3-(4-bromophenoxy)-6-methylpyridazine
2776522
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents