accessibility menu, dialog, popup

UserName

CAS RN 368869-96-9

CH3NNOBr

CAS RN 368869-96-9

IUPAC Name: 3-(4-bromophenoxy)-6-methylpyridazine
Synonyms: 3-(4-bromophenoxy)-6-methylpyridazine
Molecular Weight (g/mol): 265.11
Molecular Formula: C11H9BrN2O
InChi Key: LZEJOINDIKGWQJ-UHFFFAOYSA-N
SMILES: CC1=NN=C(OC2=CC=C(Br)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)