missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Biphenylyl)-N-Fmoc-D-alanine, 95%

Catalog No. AAH5210806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5210806 5 g
AAH5210803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5210806 Supplier Thermo Scientific Chemicals Supplier No. H5210806
Only null left

CAS: 205526-38-1 | C30H25NO4 | 463.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 205526-38-1
Molecular Formula C30H25NO4
Molecular Weight (g/mol) 463.53
MDL Number MFCD00191199
InChI Key VSGACONKQRJFGX-UHFFFAOYNA-N
Synonym fmoc-d-4,4'-biphenylalanine, fmoc-d-bip-oh, fmoc-d-bip 4,4'-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1,1'-biphenyl-4-yl propanoic acid, fmoc-4-biphenyl-d-ala, fmoc-l-phe 4-ph-oh, fmoc-bip-oh, 2r-3-1,1'-biphenyl-4-yl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, ambotzfaa1664, fmoc-d-bip 4,4atrade mark-oh
PubChem CID 7019142
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-phenylphenyl)propanoic acid
SMILES OC(=O)C(CC1=CC=C(C=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 5 g
Formula Weight 463.52
Percent Purity 95%
Chemical Name or Material 3-(4-Biphenylyl)-N-Fmoc-D-alanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only