accessibility menu, dialog, popup

UserName

OOHNHOO

CAS RN 205526-38-1

IUPAC Name: 3-{[1,1'-biphenyl]-4-yl}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: 3-{[1,1'-biphenyl]-4-yl}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 463.53
Molecular Formula: C30H25NO4
InChi Key: VSGACONKQRJFGX-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=C(C=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results