missing translation for 'onlineSavingsMsg'
Learn More

3,4-Benzopyrene (purified by sublimation) 95.0+%, TCI America™

Catalog No. B0085100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0085100MG 100 mg
B00851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0085100MG Supplier TCI America Supplier No. B0085100MG

Chemical Identifiers

CAS 50-32-8
Molecular Formula C20H12
Molecular Weight (g/mol) 252.32
MDL Number MFCD00003602
InChI Key FMMWHPNWAFZXNH-UHFFFAOYSA-N
Synonym benzo a pyrene, 3,4-benzopyrene, benzo pqr tetraphene, 3,4-benzpyrene, benzpyrene, benzo def chrysene, 6,7-benzopyrene, benz a pyrene, 3,4-benz a pyrene
PubChem CID 2336
ChEBI CHEBI:29865
IUPAC Name pentacyclo[10.6.2.0²,⁷.0⁹,¹⁹.0¹⁶,²⁰]icosa-1(18),2,4,6,8,10,12,14,16,19-decaene
SMILES C1=CC=C2C(=C1)C=C1C=CC3=CC=CC4=CC=C2C1=C34

Specifications

Melting Point 176°C
Boiling Point 496°C
Quantity 100 mg
UN Number 3077
Formula Weight 252.32
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Benzopyrene (purified by sublimation)
RTECSNumber DJ3675000
TSCA Yes