missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione, 98+%

Catalog No. p-7046156
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1332603 1 g
AAL1332606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1332603 Supplier Thermo Scientific Chemicals Supplier No. L1332603
Only null left

CAS: 20007-72-1 | C11H14O2 | 178.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20007-72-1
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00001687
InChI Key DNHDRUMZDHWHKG-UHFFFAOYNA-N
Synonym wieland-miescher ketone, 9-methyl-delta-5 10-octaline-1,6-dione, 9-methyl-5 10-octaline-1,6-dione, 1,6 2h,7h-naphthalenedione, 3,4,8,8a-tetrahydro-8a-methyl, 8a-methyl-3,4,8,8a-tetrahydro-1,6 2h,7h-naphthalenedione, wieland-michler ketone, wieland-miescher-keton, miescher-wieland ketone, wieland cmiescher ketone, 9-s-methyl-.delta.-5 10-octalin-1,6-dione
PubChem CID 89262
IUPAC Name 8a-methyl-3,4,7,8-tetrahydro-2H-naphthalene-1,6-dione
SMILES CC12CCC(=O)C=C1CCCC2=O

Specifications

Melting Point 47°C to 50°C
Quantity 1 g
Beilstein 3201286
Formula Weight 178.23
Percent Purity ≥98%
Chemical Name or Material (+/-)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione
EINECSNumber 243-463-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only