missing translation for 'onlineSavingsMsg'
Learn More

3,4,6-Tri-O-benzoyl-D-glucal, 97%

Catalog No. AAH6063703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6063703 1 g
AAH6063706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6063703 Supplier Thermo Scientific Chemicals Supplier No. H6063703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13322-90-2 | C27H22O7 | 458.466 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13322-90-2
Molecular Formula C27H22O7
Molecular Weight (g/mol) 458.466
MDL Number MFCD00061643
InChI Key OZFFEFRJEYIEGH-SMIHKQSGSA-N
Synonym 3,4,6-tri-o-benzoyl-d-glucal, tri-o-benzoyl-d-glucal, 1,2-dideoxy-d-arabino-1-hexenopyranose 3,4,6-tribenzoate, 2r,3s,4r-3,4-bis benzoyloxy-3,4-dihydro-2h-pyran-2-yl methyl benzoate
PubChem CID 11134283
IUPAC Name [(2R,3S,4R)-3,4-dibenzoyloxy-3,4-dihydro-2H-pyran-2-yl]methyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCC2C(C(C=CO2)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4

Specifications

Quantity 1 g
Formula Weight 458.46
Percent Purity 97%
Chemical Name or Material 3,4,6-Tri-O-benzoyl-D-glucal
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only