accessibility menu, dialog, popup

UserName

CAS RN 13322-90-2

IUPAC Name: [3,4-bis(benzoyloxy)-3,4-dihydro-2H-pyran-2-yl]methyl benzoate
Synonyms: [3,4-bis(benzoyloxy)-3,4-dihydro-2H-pyran-2-yl]methyl benzoate
Molecular Weight (g/mol): 458.47
Molecular Formula: C27H22O7
InChi Key: OZFFEFRJEYIEGH-UHFFFAOYNA-N
SMILES: O=C(OCC1OC=CC(OC(=O)C2=CC=CC=C2)C1OC(=O)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results