missing translation for 'onlineSavingsMsg'
Learn More

3,4,6-Tri-O-acetyl-2-deoxy-D-glucopyranose 98.0+%, TCI America™

Catalog No. T1931100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1931100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1931100MG Supplier TCI America Supplier No. T1931100MG
Only null left

Chemical Identifiers

CAS 69503-94-2
Molecular Formula C12H18O8
Molecular Weight (g/mol) 290.268
MDL Number MFCD06797168
InChI Key KQMOFVGDOWGPPD-QFEGIVONSA-N
Synonym 2-Deoxy-D-glucopyranose 3,4,6-Triacetate
PubChem CID 11748278
IUPAC Name [(2R,3S,4R)-3,4-diacetyloxy-6-hydroxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(CC(O1)O)OC(=O)C)OC(=O)C

Specifications

Melting Point 106°C
Color White
Quantity 100 mg
Formula Weight 290.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,6-Tri-O-acetyl-2-deoxy-D-glucopyranose
TSCA No
Recommended Storage Refrigerator