missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Tris(benzyloxy)benzyl Alcohol 98.0+%, TCI America™

Catalog No. t18405g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18405G Supplier TCI America Supplier No. T18405G

Chemical Identifiers

CAS 79831-88-2
Molecular Formula C28H26O4
Molecular Weight (g/mol) 426.512
MDL Number MFCD02093503
InChI Key FWPHSYJOSSBILN-UHFFFAOYSA-N
Synonym 3,4,5-Tribenzyloxybenzyl Alcohol
PubChem CID 11407668
IUPAC Name [3,4,5-tris(phenylmethoxy)phenyl]methanol
SMILES C1=CC=C(C=C1)COC2=CC(=CC(=C2OCC3=CC=CC=C3)OCC4=CC=CC=C4)CO

Specifications

Melting Point 107°C
Color White
Quantity 5 g
Formula Weight 426.51
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Tris(benzyloxy)benzyl Alcohol