missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Tris(benzyloxy)benzoic Acid 98.0+%, TCI America™

Catalog No. p-7136993
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18695G 5 g
T186925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18695G Supplier TCI America Supplier No. T18695G

Chemical Identifiers

CAS 1486-48-2
Molecular Formula C28H24O5
Molecular Weight (g/mol) 440.495
MDL Number MFCD00825832
InChI Key NRCGMEJBEDWPMG-UHFFFAOYSA-N
Synonym 3,4,5-Tribenzyloxybenzoic Acid
PubChem CID 11133969
IUPAC Name 3,4,5-tris(phenylmethoxy)benzoic acid
SMILES C1=CC=C(C=C1)COC2=CC(=CC(=C2OCC3=CC=CC=C3)OCC4=CC=CC=C4)C(=O)O

Specifications

Melting Point 196°C
Color White
Quantity 5 g
Formula Weight 440.5
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Tris(benzyloxy)benzoic Acid