accessibility menu, dialog, popup

UserName

CAS RN 1486-48-2

IUPAC Name: 3,4,5-tris(benzyloxy)benzoic acid
Synonyms: 3,4,5-tris(benzyloxy)benzoic acid
Molecular Weight (g/mol): 440.5
Molecular Formula: C28H24O5
InChi Key: NRCGMEJBEDWPMG-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(OCC2=CC=CC=C2)=C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results