missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethylphenol, 97%

Catalog No. AAL0291114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0291114 25 g
AAL0291103 1 g
AAL0291106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0291114 Supplier Thermo Scientific Chemicals Supplier No. L0291114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 527-54-8 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 527-54-8
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002305
InChI Key FDQQNNZKEJIHMS-UHFFFAOYSA-N
PubChem CID 10696
ChEBI CHEBI:38896
IUPAC Name 3,4,5-trimethylphenol
SMILES CC1=CC(=CC(=C1C)C)O

Specifications

Melting Point 104°C to 108°C
Odor Phenol-like
Quantity 25 g
UN Number UN2430
Beilstein 1907246
Synonym phenol, 3,4,5-trimethyl, 3,4,5-hemimellitenol, 1-hydroxy-3,4,5-trimethylbenzene, 5-hydroxy-1,2,3-trimethylbenzene, phenol,3,4,5-trimethyl, 3,5-hemimellitenol, 3,5-trimethylphenol, 3,4,5-trimethyl phenol, 3,4,5-trimethyl-phenol, acmc-1bhm2
Formula Weight 136.2
Percent Purity 97%
Chemical Name or Material 3,4,5-Trimethylphenol
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
EINECSNumber 208-418-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only