missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxytoluene 98.0+%, TCI America™

Catalog No. p-7136673
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T123625G 25 g
T1236500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T123625G Supplier TCI America Supplier No. T123625G
Only null left

Chemical Identifiers

CAS 6443-69-2
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD00008397
InChI Key KCIZTNZGSBSSRM-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxytoluene, benzene, 1,2,3-trimethoxy-5-methyl, 5-methylpyrogallol trimethyl ether, 1,2,3-trimethoxy-5-methyl-benzene, toluene, 3,4,5-trimethoxy, 1,2,3-trimethoxy-5-methylbenzol, pubchem13665, acmc-209nm4, 3,4,5-trimethoxy-toluene, 3,4,5-trimethoxy toluene
PubChem CID 80922
ChEBI CHEBI:81354
IUPAC Name 1,2,3-trimethoxy-5-methylbenzene
SMILES CC1=CC(=C(C(=C1)OC)OC)OC

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 182.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3,4,5-Trimethoxytoluene
EINECSNumber (3)-4659
TSCA No