missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxytoluene, 98%

Catalog No. p-7022426
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1520106 5 g
AAA1520114 25 g
AAA1520118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1520106 Supplier Thermo Scientific Chemicals Supplier No. A1520106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6443-69-2 | C10H14O3 | 182.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6443-69-2
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD00008397
InChI Key KCIZTNZGSBSSRM-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxytoluene, benzene, 1,2,3-trimethoxy-5-methyl, 5-methylpyrogallol trimethyl ether, 1,2,3-trimethoxy-5-methyl-benzene, toluene, 3,4,5-trimethoxy, 1,2,3-trimethoxy-5-methylbenzol, pubchem13665, acmc-209nm4, 3,4,5-trimethoxy-toluene, 3,4,5-trimethoxy toluene
PubChem CID 80922
ChEBI CHEBI:81354
IUPAC Name 1,2,3-trimethoxy-5-methylbenzene
SMILES CC1=CC(=C(C(=C1)OC)OC)OC

Specifications

Melting Point 34°C to 38°C
Density 1.082
Boiling Point 115°C to 116°C (4 mmHg)
Flash Point 74°C (165°F)
Refractive Index 1.523
Quantity 5 g
Beilstein 1953791
Formula Weight 182.22
Percent Purity 98%
Chemical Name or Material 3,4,5-Trimethoxytoluene
Hazard Category Warning
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 229-239-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only