missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. T28371G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T28371G Supplier TCI America Supplier No. T28371G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 182163-96-8
Molecular Formula C9H13BO5
Molecular Weight (g/mol) 212.008
MDL Number MFCD01075676
InChI Key RULQUTYJXDLRFL-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxybenzeneboronic acid, 3,4,5-trimethoxyphenyl boronic acid, 3,4,5-trimethoxyphenyl boranediol, 3,4,5-trimethoxylphenylboronic acid, boronic acid, 3,4,5-trimethoxyphenyl, pubchem7894, acmc-1c5iu, ksc174i9j, 3,4,5-trimethoxy phenylboronic acid
PubChem CID 2734390
IUPAC Name (3,4,5-trimethoxyphenyl)boronic acid
SMILES B(C1=CC(=C(C(=C1)OC)OC)OC)(O)O

Specifications

Melting Point 247°C
Color White-Yellow
Quantity 1 g
Formula Weight 212.01
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trimethoxyphenylboronic Acid (contains varying amounts of Anhydride)