accessibility menu, dialog, popup

UserName

H3COOH3COCH3BOHOH

CAS RN 182163-96-8

IUPAC Name: (3,4,5-trimethoxyphenyl)boronic acid
Synonyms: 3,4,5-trimethoxyphenylboronic acid
Molecular Weight (g/mol): 212.01
Molecular Formula: C9H13BO5
InChi Key: RULQUTYJXDLRFL-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1OC)B(O)O
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results