missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. T11055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11055G 5 g
T110525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T11055G Supplier TCI America Supplier No. T11055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 951-82-6
Molecular Formula C11H14O5
Molecular Weight (g/mol) 226.228
MDL Number MFCD00004336
InChI Key DDSJXCGGOXKGSJ-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxyphenylacetic acid, 2-3,4,5-trimethoxyphenyl acetic acid, 3,4,5-trimethoxyphenyl acetic acid, benzeneacetic acid, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenylaceticacid, pubchem14015, acmc-209rv8, ksc487g4j, 3,4,5-trimethoxybenzeneacetic acid, benzeneacetic acid,4,5-trimethoxy
PubChem CID 70372
IUPAC Name 2-(3,4,5-trimethoxyphenyl)acetic acid
SMILES COC1=CC(=CC(=C1OC)OC)CC(=O)O

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 5 g
Formula Weight 226.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trimethoxyphenylacetic Acid