Learn More
3,4,5-Trimethoxyphenylacetic acid, 99%
CAS: 951-82-6 | C11H14O5 | 226.228 g/mol
Supplier: Thermo Scientific Chemicals B2113614
Description
It is used in the synthesis of 3,4-Diaryl-2(5H)-furanones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the synthesis of 3,4-Diaryl-2(5H)-furanones.
Solubility
Soluble in water. Solubility in methanol is almost transparent.
Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 3,4,5-Trimethoxyphenylacetic acid | |
| 118°C to 122°C | |
| MFCD00004336 | |
| 2697844 | |
| Soluble in water. Solubility in methanol is almost transparent. | |
| COC1=CC(=CC(=C1OC)OC)CC(=O)O | |
| 226.228 | |
| 226.23 |
| 951-82-6 | |
| C11H14O5 | |
| 25 g | |
| 3,4,5-trimethoxyphenylacetic acid, 2-3,4,5-trimethoxyphenyl acetic acid, 3,4,5-trimethoxyphenyl acetic acid, benzeneacetic acid, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenylaceticacid, pubchem14015, acmc-209rv8, ksc487g4j, 3,4,5-trimethoxybenzeneacetic acid, benzeneacetic acid,4,5-trimethoxy | |
| DDSJXCGGOXKGSJ-UHFFFAOYSA-N | |
| 2-(3,4,5-trimethoxyphenyl)acetic acid | |
| 70372 | |
| 99% |
Chemical Identifiers
| 951-82-6 | |
| 226.228 | |
| DDSJXCGGOXKGSJ-UHFFFAOYSA-N | |
| 70372 | |
| COC1=CC(=CC(=C1OC)OC)CC(=O)O |
| C11H14O5 | |
| MFCD00004336 | |
| 3,4,5-trimethoxyphenylacetic acid, 2-3,4,5-trimethoxyphenyl acetic acid, 3,4,5-trimethoxyphenyl acetic acid, benzeneacetic acid, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenylaceticacid, pubchem14015, acmc-209rv8, ksc487g4j, 3,4,5-trimethoxybenzeneacetic acid, benzeneacetic acid,4,5-trimethoxy | |
| 2-(3,4,5-trimethoxyphenyl)acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 213-456-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only