missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyphenylacetic acid, 99%

Catalog No. AAB2113622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2113622 100 g
AAB2113606 5 g
AAB2113614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2113622 Supplier Thermo Scientific Chemicals Supplier No. B2113622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 951-82-6 | C11H14O5 | 226.228 g/mol

It is used in the synthesis of 3,4-Diaryl-2(5H)-furanones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of 3,4-Diaryl-2(5H)-furanones.

Solubility
Soluble in water. Solubility in methanol is almost transparent.

Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 951-82-6
Molecular Formula C11H14O5
Molecular Weight (g/mol) 226.228
MDL Number MFCD00004336
InChI Key DDSJXCGGOXKGSJ-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxyphenylacetic acid, 2-3,4,5-trimethoxyphenyl acetic acid, 3,4,5-trimethoxyphenyl acetic acid, benzeneacetic acid, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenylaceticacid, pubchem14015, acmc-209rv8, ksc487g4j, 3,4,5-trimethoxybenzeneacetic acid, benzeneacetic acid,4,5-trimethoxy
PubChem CID 70372
IUPAC Name 2-(3,4,5-trimethoxyphenyl)acetic acid
SMILES COC1=CC(=CC(=C1OC)OC)CC(=O)O

Specifications

Melting Point 118°C to 122°C
Quantity 100 g
Beilstein 2697844
Solubility Information Soluble in water. Solubility in methanol is almost transparent.
Formula Weight 226.23
Percent Purity 99%
Chemical Name or Material 3,4,5-Trimethoxyphenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-456-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only