missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxycinnamic acid, predominantly trans, 99%

CAS: 90-50-6 | C12H14O5 | 238.239 g/mol

Supplier:  Thermo Scientific Chemicals A1276722

 View more versions of this product

Catalog No. AAA1276722

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,4,5-Trimethoxycinnamic acid
90-50-6
C12H14O5
100 g
3,4,5-trimethoxycinnamic acid, o-methylsinapic acid, 3,4,5-trimethoxyphenylacrylic acid, 3-3,4,5-trimethoxyphenyl acrylic acid, 3-3,4,5-trimethoxyphenyl-2-propenoic acid, 3,4,5-trimethoxy cinnamic acid, sinapic acid methyl ether, e-3-3,4,5-trimethoxyphenyl acrylic acid, cinnamic acid, 3,4,5-trimethoxy, 2e-3-3,4,5-trimethoxyphenyl acrylic acid
COC1=CC(=CC(=C1OC)OC)C=CC(=O)O
238.239
238.24
predominantly trans
126°C to 128°C
MFCD00004388
1537834
YTFVRYKNXDADBI-SNAWJCMRSA-N
(E)-3-(3,4,5-trimethoxyphenyl)prop-2-enoic acid
735755
99%

Chemical Identifiers

90-50-6
238.239
YTFVRYKNXDADBI-SNAWJCMRSA-N
735755
COC1=CC(=CC(=C1OC)OC)C=CC(=O)O
C12H14O5
MFCD00004388
3,4,5-trimethoxycinnamic acid, o-methylsinapic acid, 3,4,5-trimethoxyphenylacrylic acid, 3-3,4,5-trimethoxyphenyl acrylic acid, 3-3,4,5-trimethoxyphenyl-2-propenoic acid, 3,4,5-trimethoxy cinnamic acid, sinapic acid methyl ether, e-3-3,4,5-trimethoxyphenyl acrylic acid, cinnamic acid, 3,4,5-trimethoxy, 2e-3-3,4,5-trimethoxyphenyl acrylic acid
(E)-3-(3,4,5-trimethoxyphenyl)prop-2-enoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 201-999-8

RTECSNumber : GE0722000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only