accessibility menu, dialog, popup

UserName

CAS RN 90-50-6

IUPAC Name: (2E)-3-(3,4,5-trimethoxyphenyl)prop-2-enoic acid
Synonyms: 3,4,5-trimethoxycinnamic acid
Molecular Weight (g/mol): 238.24
Molecular Formula: C12H14O5
InChi Key: YTFVRYKNXDADBI-SNAWJCMRSA-N
SMILES: COC1=CC(\C=C\C(O)=O)=CC(OC)=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results