missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzylamine 97.0+%, TCI America™

$103.97 - $103.97

Chemical Identifiers

CAS 18638-99-8
Molecular Formula C10H15NO3
Molecular Weight (g/mol) 197.234
MDL Number MFCD00017150
InChI Key YUPUSBMJCFBHAP-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxybenzylamine, 3,4,5-trimethoxyphenyl methanamine, 1-3,4,5-trimethoxyphenyl methanamine, benzenemethanamine, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methylamine, 1-3,4,5-trimethoxyphenyl methanamine hydrochloride, pubchem16674, acmc-1c5ez, ksc495e6b, 3,4,5-trimethoxy benzylamine
PubChem CID 87736
IUPAC Name (3,4,5-trimethoxyphenyl)methanamine
SMILES COC1=CC(=CC(=C1OC)OC)CN
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T30875G
View Documents
TCI America
T30875G
5 g
Each for $103.97
Only null left
 
Description

Description

Specifications

Chemical Identifiers

18638-99-8
197.234
YUPUSBMJCFBHAP-UHFFFAOYSA-N
87736
COC1=CC(=CC(=C1OC)OC)CN
C10H15NO3
MFCD00017150
3,4,5-trimethoxybenzylamine, 3,4,5-trimethoxyphenyl methanamine, 1-3,4,5-trimethoxyphenyl methanamine, benzenemethanamine, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methylamine, 1-3,4,5-trimethoxyphenyl methanamine hydrochloride, pubchem16674, acmc-1c5ez, ksc495e6b, 3,4,5-trimethoxy benzylamine
(3,4,5-trimethoxyphenyl)methanamine

Specifications

18638-99-8
121°C
MFCD00017150
2735
YUPUSBMJCFBHAP-UHFFFAOYSA-N
(3,4,5-trimethoxyphenyl)methanamine
87736
≥97.0% (GC,T)
3,4,5-Trimethoxybenzylamine
Yellow
C10H15NO3
5 g
3,4,5-trimethoxybenzylamine, 3,4,5-trimethoxyphenyl methanamine, 1-3,4,5-trimethoxyphenyl methanamine, benzenemethanamine, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methylamine, 1-3,4,5-trimethoxyphenyl methanamine hydrochloride, pubchem16674, acmc-1c5ez, ksc495e6b, 3,4,5-trimethoxy benzylamine
COC1=CC(=CC(=C1OC)OC)CN
197.234
197.23
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents