missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzeneboronic acid, 98%

Catalog No. AAL1519103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1519103 1 g
AAL1519106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1519103 Supplier Thermo Scientific Chemicals Supplier No. L1519103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 182163-96-8 | C9H13BO5 | 212.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 182163-96-8
Molecular Formula C9H13BO5
Molecular Weight (g/mol) 212.008
MDL Number MFCD01075676
InChI Key RULQUTYJXDLRFL-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxybenzeneboronic acid, 3,4,5-trimethoxyphenyl boronic acid, 3,4,5-trimethoxyphenyl boranediol, 3,4,5-trimethoxylphenylboronic acid, boronic acid, 3,4,5-trimethoxyphenyl, pubchem7894, acmc-1c5iu, ksc174i9j, 3,4,5-trimethoxy phenylboronic acid
PubChem CID 2734390
IUPAC Name (3,4,5-trimethoxyphenyl)boronic acid
SMILES B(C1=CC(=C(C(=C1)OC)OC)OC)(O)O

Specifications

Melting Point ∼245°C
Quantity 1 g
Beilstein 8205051
Formula Weight 212.01
Percent Purity 98%
Chemical Name or Material 3,4,5-Trimethoxybenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only