Learn More
3,4,5-Trimethoxybenzaldehyde, 99%
CAS: 86-81-7 | C10H12O4 | 196.2 g/mol
$66.98 - $66.98
Chemical Identifiers
| CAS | 86-81-7 |
|---|---|
| Molecular Formula | C10H12O4 |
| Molecular Weight (g/mol) | 196.2 |
| MDL Number | MFCD00003364 |
| InChI Key | OPHQOIGEOHXOGX-UHFFFAOYSA-N |
| PubChem CID | 6858 |
| IUPAC Name | 3,4,5-trimethoxybenzaldehyde |
| SMILES | COC1=CC(=CC(=C1OC)OC)C=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC139910250
|
Thermo Scientific Chemicals
139910250 |
25 g | Glass bottle |
Each for $66.98
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 86-81-7 | |
| 196.2 | |
| OPHQOIGEOHXOGX-UHFFFAOYSA-N | |
| 3,4,5-trimethoxybenzaldehyde |
| C10H12O4 | |
| MFCD00003364 | |
| 6858 | |
| COC1=CC(=CC(=C1OC)OC)C=O |
Specifications
| 86-81-7 | |
| 100.0 | |
| Cream to White | |
| Authentic | |
| Glass bottle | |
| (CH3O)3C6H2CHO | |
| 25 g | |
| benzaldehyde, 3,4,5-trimethoxy, unii-wl86yd76n6, tmba, benzaldehyde,3,4,5-trimethoxy, 3,4,5-trimethoxy benzaldehyde, 3,4,5-trimethoxy-benzaldehyde, pubchem8266, acmc-209qbj, 3,5-trimethoxybenzaldehyde, 2.3.4-trihyroxybenzaldehyde | |
| OPHQOIGEOHXOGX-UHFFFAOYSA-N | |
| 3,4,5-trimethoxybenzaldehyde | |
| 6858 | |
| 99% | |
| 3, 4, 5-Trimethoxybenzaldehyde |
| 98.5 | |
| 73.0°C to 76.0°C | |
| 163.0°C to 165.0°C (10.0 mmHg) | |
| 98.5% min. (GC) | |
| C10H12O4 | |
| MFCD00003364 | |
| 08, 391 | |
| Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,toluene and acetone | |
| COC1=CC(=CC(=C1OC)OC)C=O | |
| 196.2 | |
| 196.2 | |
| Powder or Flakes |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 201-701-6
RUO – Research Use Only